missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, meets analytical specification of Ph.Eur

Product Code. 11994051
Click to view available options
Quantity:
1 kg
3 kg
10 kg
25 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Quantity 3 kg
IUPAC Name sodium chloride
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'