missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate, ACS Reagent, anhydrous, powder or granules, ≥99.5%, Honeywell Fluka™
ACS Reagent, anhydrous, powder or granules,≥99.5%
40.53 € - 580.75 €
Chemical Identifiers
CAS | 497-19-8 |
---|---|
Molecular Formula | CNa2O3 |
Molecular Weight (g/mol) | 105.99 |
MDL Number | MFCD00003494 |
InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
Synonym | sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda |
PubChem CID | 10340 |
ChEBI | CHEBI:29377 |
SMILES | [Na+].[Na+].[O-]C([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15602190
|
Honeywell-Fluka
222321-500G |
500 g | Poly Bottle | ||||||
15672180
|
Honeywell-Fluka
222321-1KG |
1 kg | Poly Bottle | ||||||
15682180
|
Honeywell-Fluka
222321-2.5KG |
2.5 kg | Poly Bottle | ||||||
15612190
|
Honeywell-Fluka
222321-5KG |
5 kg | Drum With Inliner | ||||||
15692180
|
Honeywell-Fluka
222321-4X2.5KG |
4 x 2.5 kg | Poly Bottle | ||||||
15662180
|
Honeywell-Fluka
222321-12KG |
12 kg | Fiberboard Box | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
497-19-8 | |
105.99 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
10340 | |
[Na+].[Na+].[O-]C([O-])=O |
CNa2O3 | |
MFCD00003494 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
CHEBI:29377 |
Specifications
851°C (literature) | |
≥90.0000% | |
Powder/Granules | |
Na2CO3 | |
4154566 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium carbonate | |
10340 | |
105.99 | |
ACS Reagent | |
ex ignited substance |
497-19-8 | |
≤100.0000% | |
CNa2O3 | |
MFCD00003494 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
[Na+].[Na+].[O-]C([O-])=O | |
105.99 | |
CHEBI:29377 | |
>99.5% | |
2.53 g/cm3 | |
Sodium carbonate, ACS Reagent, anhydrous, powder or granules, ≥99.5% |
Safety and Handling
missing translation for 'shelfLife' : 1260 days from date of manufacture
missing translation for 'einecsNumber' : 207-838-8
Recommended Storage : Room Temp