missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate, ACS Reagent, anhydrous, powder or granules, ≥99.5%, Honeywell Fluka™

Product Code. 15612190
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
12 kg
4 x 2.5 kg
Packaging:
Drum With Inliner
Fiberboard Box
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent, anhydrous, powder or granules,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Melting Point 851°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Physical Form Powder/Granules
Quantity 5 kg
Linear Formula Na2CO3
Beilstein 4154566
IUPAC Name disodium carbonate
Formula Weight 105.99
Percent Purity >99.5%
Grade ACS Reagent
Packaging Drum With Inliner
Density 2.53 g/cm3
Assay ex ignited substance
Chemical Name or Material Sodium carbonate, ACS Reagent, anhydrous, powder or granules, ≥99.5%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'