missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate, ≥99.5%, Reagent Grade, Honeywell™
Reagent Grade,≥99.5%
31.22 € - 210.48 €
Chemical Identifiers
CAS | 497-19-8 |
---|---|
Molecular Formula | CNa2O3 |
Molecular Weight (g/mol) | 105.99 |
MDL Number | MFCD00003494 |
InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
Synonym | sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda |
PubChem CID | 10340 |
ChEBI | CHEBI:29377 |
SMILES | [Na+].[Na+].[O-]C([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15648080
|
Honeywell Research Chemicals
S2127-500G |
500 g | Poly bottle | N/A | |||||
15638080
|
Honeywell Research Chemicals
S2127-1KG |
1 kg | Poly bottle | N/A | |||||
15658080
|
Honeywell Research Chemicals
S2127-5KG |
5 kg | Poly bottle | N/A | |||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
497-19-8 | |
105.99 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
10340 | |
[Na+].[Na+].[O-]C([O-])=O |
CNa2O3 | |
MFCD00003494 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
CHEBI:29377 |
Specifications
854°C | |
White | |
Solid | |
Na2CO3 | |
NONH for all modes of transport | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
[Na+].[Na+].[O-]C([O-])=O | |
105.99 | |
CHEBI:29377 | |
≥99.5% | |
2.53g/cu.cm at 20°C |
497-19-8 | |
11.3 (at 10.00g/L, 20°C) | |
CNa2O3 | |
MFCD00003494 | |
4154566 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium carbonate | |
10340 | |
105.99g/mol | |
Odorless | |
Sodium carbonate |
Safety and Handling
P305 + P351 + P338
missing translation for 'rtecsNumber' : 207-838-8
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title