missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate, ≥99.5%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Poly bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | 854°C |
Color | White |
pH | 11.3 (at 10.00g/L, 20°C) |
Physical Form | Solid |
Quantity | 500 g |
Linear Formula | Na2CO3 |
UN Number | NONH for all modes of transport |
Beilstein | 4154566 |
IUPAC Name | disodium carbonate |
Formula Weight | 105.99g/mol |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction