missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC37A1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | SLC37A1 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883065
|
Proteintech
24582-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873065
|
Proteintech
24582-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC37A1, a member of the sugar-phosphate transport family, transports glycerol-3-phosphate (G3P) between cellular compartments for its utilization in several compartment-specific biochemical pathways.Specifications
SLC37A1 | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
P57057, Q8R070 | |
224674, 54020 | |
SLC37A1 Fusion Protein Ag20180 | |
Primary | |
-20°C | |
SLC37A1 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
G 3 P permease, G 3 P transporter, G3PP, Glycerol 3 phosphate permease, SLC37A1 | |
SLC37A1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title