missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC37A1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24582-1-AP-150UL
This item is not returnable.
View return policy
Description
SLC37A1, a member of the sugar-phosphate transport family, transports glycerol-3-phosphate (G3P) between cellular compartments for its utilization in several compartment-specific biochemical pathways.Specifications
SLC37A1 | |
Polyclonal | |
Unconjugated | |
SLC37A1 | |
G 3 P permease, G 3 P transporter, G3PP, Glycerol 3 phosphate permease, SLC37A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
224674, 54020 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P57057, Q8R070 | |
SLC37A1 | |
SLC37A1 Fusion Protein Ag20180 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction