missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon Dioxide, calcined, crude, Honeywell Fluka™
for cleaning of platinum crucibles, calcined, crude
83.39 € - 410.04 €
Chemical Identifiers
CAS | 14808-60-7 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.08 |
MDL Number | MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
Synonym | silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | dioxosilane |
SMILES | O=[Si]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15671680
|
Honeywell-Fluka
18648-2.5KG |
2.5 kg |
83.39 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15691680
|
Honeywell-Fluka
18648-6X2.5KG |
6 x 2.5 kg |
410.04 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15681680
|
Honeywell-Fluka
18648-50KG |
50 kg |
211.17 €
50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
14808-60-7 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
dioxosilane |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
CHEBI:30563 | |
O=[Si]=O |
Specifications
14808-60-7 | |
SiO2 | |
NONH for all modes of transport | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
60.08g/mol |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 | |
Silicon Dioxide |
Safety and Handling
missing translation for 'einecsNumber' : 238-878-4
missing translation for 'rtecsNumber' : VV7330000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title