missing translation for 'onlineSavingsMsg'
Learn More

Silicon Dioxide, calcined, crude, Honeywell Fluka™

Product Code. 15681680
Click to view available options
Quantity:
2.5 kg
50 kg
6 x 2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

for cleaning of platinum crucibles, calcined, crude

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Quantity 50 kg
Linear Formula SiO2
UN Number NONH for all modes of transport
Formula Weight 60.08g/mol
Chemical Name or Material Silicon Dioxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'