missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sibutramine hydrochloride monohydrate, Thermo Scientific Chemicals
69.70 € - 211.00 €
Chemical Identifiers
CAS | 125494-59-9 |
---|---|
Molecular Formula | C17H29Cl2NO |
Molecular Weight (g/mol) | 334.33 |
InChI Key | KFNNPQDSPLWLCX-UHFFFAOYNA-N |
PubChem CID | 64765 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
125494-59-9 | |
KFNNPQDSPLWLCX-UHFFFAOYNA-N | |
64765 |
C17H29Cl2NO | |
334.33 | |
Sibutramine hydrochloride monohydrate |
RUO – Research Use Only