missing translation for 'onlineSavingsMsg'
Learn More

Sibutramine hydrochloride monohydrate, Thermo Scientific Chemicals

Product Code. 15771799
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125494-59-9
Molecular Formula C17H29Cl2NO
Molecular Weight (g/mol) 334.33
InChI Key KFNNPQDSPLWLCX-UHFFFAOYNA-N
PubChem CID 64765

Specifications

Quantity 1 g
Chemical Name or Material Sibutramine hydrochloride monohydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'