Learn More
Epredia™ Embedding Cassettes in Tube Packs
Hold and identify tissue samples in processing, embedding and sectioning procedures with Embedding Cassettes in Tube Packs. Compatible for use with the Epredia™ PrintMate AS™ Cassette Printer.
Brand:  Epredia™ B1000732GR
 View more versions of this product
Product Code. 12993785
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Can be used on all Epredia PrintMate automatic cassette labelers
  • Fit securely in microtome chuck adapters
  • Designed to accept metal lids (Order number 1001003, 25/pk., sold separately)
  • Made of acetal polymer with a pore style of round holes
  • Supplied in 10 tubes of 75 cassettes each
Specifications
Specifications
Round Holes
750 Cassettes/Cs.
Green
Acetal Polymer