missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGK1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | SGK1 |
---|---|
Concentration | 450 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16886145
|
Proteintech
28454-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16876145
|
Proteintech
28454-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SGK1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SGK1. This antibody recognizes human antigen. The SGK1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
SGK1 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human | |
IgG |
450 μg/mL | |
Polyclonal | |
Rabbit | |
6446 | |
Primary |