Learn More
SGK1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28454-1-AP-20UL
This item is not returnable.
View return policy
Description
The SGK1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SGK1. This antibody recognizes human antigen. The SGK1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
SGK1 | |
Polyclonal | |
Unconjugated | |
20 μL | |
6446 | |
IgG |
Western Blot, Immunohistochemistry | |
450 μg/mL | |
Rabbit | |
Primary | |
Human |