missing translation for 'onlineSavingsMsg'
Learn More

SDHD Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen SDHD
Concentration 167 μg/mL
Applications Immunofluorescence, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16844955
View Documents Promotion Details
Proteintech
26519-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16834955
View Documents Promotion Details
Proteintech
26519-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The SDHD antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human SDHD. This antibody recognizes human antigen. The SDHD antibody has been validated for the following applications: IF, ELISA analysis.

Specifications

Specifications

SDHD
Immunofluorescence, ELISA
Unconjugated
Human
IgG
167 μg/mL
Polyclonal
Rabbit
6392
Primary
Videos
SDS
Documents

Documents

Product Certifications