Learn More
SDHD Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26519-1-AP-20UL
This item is not returnable.
View return policy
Description
The SDHD antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human SDHD. This antibody recognizes human antigen. The SDHD antibody has been validated for the following applications: IF, ELISA analysis.
Specifications
SDHD | |
Polyclonal | |
Unconjugated | |
20 μL | |
6392 | |
IgG |
Immunofluorescence, ELISA | |
167 μg/mL | |
Rabbit | |
Primary | |
Human |