missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Cubis™ Balance Display and Control Units
Variety of displays and control units available
Brand: Sartorius YAC01MSA
This item is not returnable.
View return policy
Description
For use with Cubis™ balances.Specifications
Cubis™ Balance | |
Color TFT graphic display and touch screen | |
Control Unit |