missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Cubis™ Balance Display and Control Units
Variety of displays and control units available
1940.00 € - 3040.00 €
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
13169240
|
Sartorius
YAC01MSU |
Backlit black and white graphic display and tactil |
1940.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16373086
|
Sartorius
YAC01MSA |
Color TFT graphic display and touch screen |
3040.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For use with Cubis™ balances.Specifications
Cubis™ Balance | |
Control Unit |