Learn More
Hamilton™ SaltLine™ Luer Lock Syringes
Choose these syringes when a precise flow is critical for almost any application.
195.00 € - 216.00 €
Products 4
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
10491591
View Documents
Hamilton™
203260
5 mL
196.00 €
Each
10777534
View Documents
Hamilton™
203270
10 mL
195.00 €
Each
11522495
View Documents
Hamilton™
203240
200.00 €
Each
11562455
View Documents
Hamilton™
203220
250 μL
216.00 €
Each
Description
Description
  • PTFE luer lock termination is extremely common as it provides the most flexible options for connection of tubing
  • Available in a range of volumes
  • Applications include microfluidics, drug/nutritional infusions, electrospinning, mass spectrometer calibration, cell feeding and more