Learn More
Hamilton™ SaltLine™ Luer Lock Syringes
Choose these syringes when a precise flow is critical for almost any application.
Brand: Hamilton™ 203270
This item is not returnable.
View return policy
Description
- PTFE luer lock termination is extremely common as it provides the most flexible options for connection of tubing
- Available in a range of volumes
- Applications include microfluidics, drug/nutritional infusions, electrospinning, mass spectrometer calibration, cell feeding and more
Specifications
Not Autoclavable | |
SaltLine Luer Lock Gastight Syringe | |
Laboratory Syringe Pumps |
Borosilicate Glass, PTFE, Stainless Steel, UHMWPE | |
10 mL | |
Luer Lock |