missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S,S)-DIPAMP, 97%, Thermo Scientific Chemicals
78.70 € - 401.00 €
Chemical Identifiers
CAS | 97858-62-3 |
---|---|
Molecular Formula | C28H28O2P2 |
Molecular Weight (g/mol) | 458.48 |
MDL Number | MFCD05863547 |
InChI Key | QKZWXPLBVCKXNQ-UHFFFAOYNA-N |
Synonym | 1,2-bis s-2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp, unii-hs6f5ew3u6, hs6f5ew3u6, ethylenebis s-o-anisyl phenyl phosphine, s,s-+-1,2-bis 2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp; s,s-ethylenebis 2-methoxyphenyl phenylphosphine, s-2-methoxyphenyl 2-s-2-methoxyphenyl phenyl phosphanyl ethyl phenylphosphane |
PubChem CID | 11419748 |
IUPAC Name | (S)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane |
SMILES | COC1=CC=CC=C1P(CCP(C1=CC=CC=C1)C1=CC=CC=C1OC)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11357569
|
Thermo Scientific Alfa Aesar
H28037.MC |
100 mg |
78.70 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15423817
|
Thermo Scientific Alfa Aesar
H28037.MD |
250 mg | |||||||
11377569
|
Thermo Scientific Alfa Aesar
H28037.03 |
1 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
97858-62-3 | |
458.48 | |
QKZWXPLBVCKXNQ-UHFFFAOYNA-N | |
11419748 | |
COC1=CC=CC=C1P(CCP(C1=CC=CC=C1)C1=CC=CC=C1OC)C1=CC=CC=C1 |
C28H28O2P2 | |
MFCD05863547 | |
1,2-bis s-2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp, unii-hs6f5ew3u6, hs6f5ew3u6, ethylenebis s-o-anisyl phenyl phosphine, s,s-+-1,2-bis 2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp; s,s-ethylenebis 2-methoxyphenyl phenylphosphine, s-2-methoxyphenyl 2-s-2-methoxyphenyl phenyl phosphanyl ethyl phenylphosphane | |
(S)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane |
Specifications
97858-62-3 | |
C28H28O2P2 | |
1,2-bis s-2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp, unii-hs6f5ew3u6, hs6f5ew3u6, ethylenebis s-o-anisyl phenyl phosphine, s,s-+-1,2-bis 2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp; s,s-ethylenebis 2-methoxyphenyl phenylphosphine, s-2-methoxyphenyl 2-s-2-methoxyphenyl phenyl phosphanyl ethyl phenylphosphane | |
+84° (c=1 in Chloroform) | |
(S)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane | |
11419748 | |
97% |
101°C to 105°C | |
MFCD05863547 | |
QKZWXPLBVCKXNQ-UHFFFAOYNA-N | |
COC1=CC=CC=C1P(CCP(C1=CC=CC=C1)C1=CC=CC=C1OC)C1=CC=CC=C1 | |
458.48 | |
458.47 | |
(S,S)-DIPAMP |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only