missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-DIPAMP, 97%, Thermo Scientific Chemicals

Product Code. 11377569
Click to view available options
Quantity:
100 mg
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97858-62-3
Molecular Formula C28H28O2P2
Molecular Weight (g/mol) 458.48
MDL Number MFCD05863547
InChI Key QKZWXPLBVCKXNQ-UHFFFAOYNA-N
Synonym 1,2-bis s-2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp, unii-hs6f5ew3u6, hs6f5ew3u6, ethylenebis s-o-anisyl phenyl phosphine, s,s-+-1,2-bis 2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp; s,s-ethylenebis 2-methoxyphenyl phenylphosphine, s-2-methoxyphenyl 2-s-2-methoxyphenyl phenyl phosphanyl ethyl phenylphosphane
PubChem CID 11419748
IUPAC Name (S)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane
SMILES COC1=CC=CC=C1P(CCP(C1=CC=CC=C1)C1=CC=CC=C1OC)C1=CC=CC=C1

Specifications

Melting Point 101°C to 105°C
Quantity 1 g
Optical Rotation +84° (c=1 in Chloroform)
Formula Weight 458.47
Percent Purity 97%
Chemical Name or Material (S,S)-DIPAMP

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'