missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2-(Benzyloxycarbonylamino)butyric acid, 98%, Thermo Scientific Chemicals
46.90 € - 667.00 €
Chemical Identifiers
CAS | 42918-86-5 |
---|---|
Molecular Formula | C12H15NO4 |
Molecular Weight (g/mol) | 237.255 |
MDL Number | MFCD00077006 |
InChI Key | SZQMTCSQWUYUML-JTQLQIEISA-N |
Synonym | z-abu-oh, s-2-benzyloxy carbonyl amino butanoic acid, 2s-2-benzyloxy carbonyl amino butanoic acid, z-2-abu-oh, s-2-bezyloxycarbonylamino butanoic acid, s-2-benzyloxycarbonylamino butanoic acid, butanoic acid, 2-phenylmethoxy carbonyl amino-, 2s, ambotzzaa1200, pubchem18943, s-2-cbz-amino butyricacid |
PubChem CID | 7349998 |
IUPAC Name | (2S)-2-(phenylmethoxycarbonylamino)butanoic acid |
SMILES | CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15468928
|
Thermo Scientific Alfa Aesar
H62426.03 |
1 g | N/A | ||||||
15478928
|
Thermo Scientific Alfa Aesar
H62426.06 |
5 g | N/A | ||||||
15488928
|
Thermo Scientific Alfa Aesar
H62426.14 |
25 g |
667.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
42918-86-5 | |
237.255 | |
SZQMTCSQWUYUML-JTQLQIEISA-N | |
7349998 | |
CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1 |
C12H15NO4 | |
MFCD00077006 | |
z-abu-oh, s-2-benzyloxy carbonyl amino butanoic acid, 2s-2-benzyloxy carbonyl amino butanoic acid, z-2-abu-oh, s-2-bezyloxycarbonylamino butanoic acid, s-2-benzyloxycarbonylamino butanoic acid, butanoic acid, 2-phenylmethoxy carbonyl amino-, 2s, ambotzzaa1200, pubchem18943, s-2-cbz-amino butyricacid | |
(2S)-2-(phenylmethoxycarbonylamino)butanoic acid |
Specifications
42918-86-5 | |
MFCD00077006 | |
SZQMTCSQWUYUML-JTQLQIEISA-N | |
(2S)-2-(phenylmethoxycarbonylamino)butanoic acid | |
7349998 | |
98% |
C12H15NO4 | |
z-abu-oh, s-2-benzyloxy carbonyl amino butanoic acid, 2s-2-benzyloxy carbonyl amino butanoic acid, z-2-abu-oh, s-2-bezyloxycarbonylamino butanoic acid, s-2-benzyloxycarbonylamino butanoic acid, butanoic acid, 2-phenylmethoxy carbonyl amino-, 2s, ambotzzaa1200, pubchem18943, s-2-cbz-amino butyricacid | |
CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1 | |
237.255 | |
237.25 | |
(S)-2-(Benzyloxycarbonylamino)butyric acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only