missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(Benzyloxycarbonylamino)butyric acid, 98%, Thermo Scientific Chemicals

Product Code. 15468928
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42918-86-5
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00077006
InChI Key SZQMTCSQWUYUML-JTQLQIEISA-N
Synonym z-abu-oh, s-2-benzyloxy carbonyl amino butanoic acid, 2s-2-benzyloxy carbonyl amino butanoic acid, z-2-abu-oh, s-2-bezyloxycarbonylamino butanoic acid, s-2-benzyloxycarbonylamino butanoic acid, butanoic acid, 2-phenylmethoxy carbonyl amino-, 2s, ambotzzaa1200, pubchem18943, s-2-cbz-amino butyricacid
PubChem CID 7349998
IUPAC Name (2S)-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Formula Weight 237.25
Percent Purity 98%
Quantity 1 g
Chemical Name or Material (S)-2-(Benzyloxycarbonylamino)butyric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'