Learn More
RB1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10048-2-IG-20UL
 View more versions of this product
Product Code. 16886193
  • 145.00 € / 20µL
Explore more special offers
Description
Description
This antibody is a rabbit polyclonal antibody raised against human RB1 fusion protein.
Rb is a tumor suppressor nuclear phosphoprotein capable of binding to DNA. It is phosphorylated on serine and threonine, but not on tyrosine residues. It forms a complex with SV40 large T antigen, adenovirus E1A, and human papilloma virus-16 E. Rb protein may act by regulating transcription and loss of its function leads to uncontrolled cell growth. Aberrations in the RB gene have been implicated in cancers of breast, colon, prostate, kidney, nasopharynx, and leukemia. Phosphorylation of Rb at S608 depends on CDK4.
Specifications
Specifications
RB1
Polyclonal
Unconjugated
RB1
OSRC, p105 Rb, pp110, pRb, RB, RB1, retinoblastoma 1
Rabbit
Protein A
RUO
19645, 5925
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P06400, P13405
RB1
RB1 Fusion Protein Ag0006
20 μL
Primary
Human, Mouse