missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Quinaldine red, pure
69.75 € - 751.74 €
Chemical Identifiers
CAS | 117-92-0 |
---|---|
Molecular Formula | C21H23IN2 |
Molecular Weight (g/mol) | 430.33 |
MDL Number | MFCD00011968 |
InChI Key | JOLANDVPGMEGLK-UHFFFAOYSA-M |
Synonym | 2-[4-(Dimethylamino)styryl]-1-ethylquinolinium iodide |
PubChem CID | 5702759 |
SMILES | [I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10200023
|
Thermo Scientific Chemicals
377510050 |
5 g | Glass bottle |
69.75 €
5g |
In Stock
Log in to see stock. |
||||
10620545
|
Thermo Scientific Chemicals
377510250 |
25 g | Glass bottle |
235.17 €
25g |
In Stock
Log in to see stock. |
||||
10638994
|
Thermo Scientific Chemicals
377511000 |
100 g | Glass bottle |
751.74 €
100g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
117-92-0 | |
430.33 | |
JOLANDVPGMEGLK-UHFFFAOYSA-M | |
5702759 |
C21H23IN2 | |
MFCD00011968 | |
2-[4-(Dimethylamino)styryl]-1-ethylquinolinium iodide | |
[I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C |
Specifications
117-92-0 | |
MFCD00011968 | |
JOLANDVPGMEGLK-UHFFFAOYSA-M | |
[I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C | |
430.33 | |
430.32 | |
Complies | |
Powder |
C21H23IN2 | |
2-[4-(Dimethylamino)styryl]-1-ethylquinolinium iodide | |
Authentic | |
2-{2-[4-(dimethylamino)phenyl]ethenyl}-1-ethylquinolin-1-ium iodide | |
5702759 | |
Pure | |
Black/Green or Blue to Gray | |
Quinaldine red, Pure |
Safety and Handling
Product Identifier
- Quinaldine Red
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title