missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Quinaldine red, pure
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
Infrared Spectrum | Authentic |
IUPAC Name | 4-[(E)-2-(1-ethylquinolin-1-ium-2-yl)ethenyl]-N,N-dimethylaniline;iodide |
Formula Weight | 430.32 |
Grade | Pure |
Packaging | Glass bottle |
UV Spectrometry | Complies |
Color | Black/Green or Blue to Gray |
Quantity | 25 g |
Physical Form | Powder |
Chemical Name or Material | Quinaldine red, Pure |
Safety and Handling
Product Identifier
- Quinaldine Red
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction