Learn More
Pyrazine-2-thiocarboxamide, 97%, Thermo Scientific Chemicals
44.20 € - 376.00 €
Chemical Identifiers
CAS | 4604-72-2 |
---|---|
Molecular Formula | C5H5N3S |
Molecular Weight (g/mol) | 139.176 |
MDL Number | MFCD00173663 |
InChI Key | LIURPUMROGYCLW-UHFFFAOYSA-N |
Synonym | thiopyrazinamide, pyrazinecarbothioamide, pyrazinecarboxamide, thio, pyrazine-2-thiocarboxamide, 2-pyrazinecarbothioamide, pyrazinecarboxylic acid thioamide, 2-pyrazinethiocarboxamide, pyrazine carbothioamide, pyrazine thiocarboxamide, acmc-20ajt5 |
PubChem CID | 2797467 |
IUPAC Name | pyrazine-2-carbothioamide |
SMILES | C1=CN=C(C=N1)C(=S)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15433537
|
Thermo Scientific Alfa Aesar
H26750.MD |
250 mg |
44.20 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346809
|
Thermo Scientific Alfa Aesar
H26750.03 |
1 g | N/A | ||||||
11356809
|
Thermo Scientific Alfa Aesar
H26750.06 |
5 g |
376.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in organic solvents, ethanol, DMSO.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
4604-72-2 | |
139.176 | |
LIURPUMROGYCLW-UHFFFAOYSA-N | |
2797467 | |
C1=CN=C(C=N1)C(=S)N |
C5H5N3S | |
MFCD00173663 | |
thiopyrazinamide, pyrazinecarbothioamide, pyrazinecarboxamide, thio, pyrazine-2-thiocarboxamide, 2-pyrazinecarbothioamide, pyrazinecarboxylic acid thioamide, 2-pyrazinethiocarboxamide, pyrazine carbothioamide, pyrazine thiocarboxamide, acmc-20ajt5 | |
pyrazine-2-carbothioamide |
Specifications
4604-72-2 | |
Odorless | |
MFCD00173663 | |
Soluble in organic solvents,ethanol,DMSO. | |
C1=CN=C(C=N1)C(=S)N | |
139.176 | |
139.17 | |
Pyrazine-2-thiocarboxamide |
198°C to 200°C | |
C5H5N3S | |
thiopyrazinamide, pyrazinecarbothioamide, pyrazinecarboxamide, thio, pyrazine-2-thiocarboxamide, 2-pyrazinecarbothioamide, pyrazinecarboxylic acid thioamide, 2-pyrazinethiocarboxamide, pyrazine carbothioamide, pyrazine thiocarboxamide, acmc-20ajt5 | |
LIURPUMROGYCLW-UHFFFAOYSA-N | |
pyrazine-2-carbothioamide | |
2797467 | |
97% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
H302-H315-H317-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only