missing translation for 'onlineSavingsMsg'
Learn More

Pyrazine-2-thiocarboxamide, 97%, Thermo Scientific Chemicals

Product Code. 11346809
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in organic solvents, ethanol, DMSO.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4604-72-2
Molecular Formula C5H5N3S
Molecular Weight (g/mol) 139.176
MDL Number MFCD00173663
InChI Key LIURPUMROGYCLW-UHFFFAOYSA-N
Synonym thiopyrazinamide, pyrazinecarbothioamide, pyrazinecarboxamide, thio, pyrazine-2-thiocarboxamide, 2-pyrazinecarbothioamide, pyrazinecarboxylic acid thioamide, 2-pyrazinethiocarboxamide, pyrazine carbothioamide, pyrazine thiocarboxamide, acmc-20ajt5
PubChem CID 2797467
IUPAC Name pyrazine-2-carbothioamide
SMILES C1=CN=C(C=N1)C(=S)N

Specifications

Melting Point 198°C to 200°C
Odor Odorless
Quantity 1 g
Solubility Information Soluble in organic solvents,ethanol,DMSO.
Formula Weight 139.17
Percent Purity 97%
Chemical Name or Material Pyrazine-2-thiocarboxamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'