Learn More
Buerkle™ PVC Standard Bung
Laboratory standard bung made of PVC with good chemical resistance against acids and alkalis.
0.95 € - 6.85 €
Specifications
Color | Red |
---|
Alert:
Attention! Make sure to pay attention to the List of Chemical Resistance!
Product Code | Brand | Diameter (Metric) Top | Diameter (Metric) Bottom | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Top | Diameter (Metric) Bottom | Height (Metric) | Price | Quantity & Availability | |||
17210644
|
Buerkle™
9763-0913 |
13 mm | 9 mm | 20 mm |
0.95 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||
17250644
|
Buerkle™
9763-1924 |
24 mm | 19 mm | 25 mm |
4.35 €
Each |
In Stock
Log in to see stock. |
|||
17220644
|
Buerkle™
9763-1115 |
15 mm | 11 mm | 20 mm |
1.15 €
Each |
In Stock
Log in to see stock. |
|||
17260644
|
Buerkle™
9763-2227 |
27 mm | 22 mm | 24 mm |
4.85 €
Each |
In Stock
Log in to see stock. |
|||
17230644
|
Buerkle™
9763-1418 |
18 mm | 14 mm | 20 mm |
3.55 €
Each |
In Stock
Log in to see stock. |
|||
17270644
|
Buerkle™
9763-2834 |
30 mm | N/A | ||||||
17240644
|
Buerkle™
9763-1621 |
21 mm | 16 mm | 24 mm |
4.20 €
Each |
In Stock
Log in to see stock. |
|||
17280644
|
Buerkle™
9763-3138 |
33 mm |
5.50 €
Each |
In Stock
Log in to see stock. |
|||||
17290644
|
Buerkle™
9763-4249 |
34 mm |
6.85 €
Each |
In Stock
Log in to see stock. |
|||||
Description
- PVC, soft, red
Specifications
Red |