Learn More
Buerkle™ PVC Standard Bung
Laboratory standard bung made of PVC with good chemical resistance against acids and alkalis.
Brand:  Buerkle™ 9763-3138
4.57 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 17280644
  • 5.50 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Alert:
Attention! Make sure to pay attention to the List of Chemical Resistance!
Description
Description
  • PVC, soft, red
Specifications
Specifications
33 mm
31 mm
38 mm
Red