Learn More
DuPont™ Proshield™ 4 Practik
Proshield™ 4 Practik suits are the ideal work suits for general maintenance in industrial manufacturing and other harmless environments.
46.11 € - 184.44 €
Specifications
Gender Unisex
Material Polyethylene
Color White
Disposable Single-use
Cuff Style Elastic
View More Specs
Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
16263512
View Documents
DuPont™
D13495537
Medium This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16253512
View Documents
DuPont™
D13495513
Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16283502
View Documents
DuPont™
D13396479
X-Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16273502
View Documents
DuPont™
D13396469
2X-Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Micro-perforated non-woven polyethylene fabric allows unrestricted passage of air and water vapour
  • Certified according to Regulation (EU) 2016/425
  • Chemical protective clothing, Category I
  • Antistatic treatment (EN 1149-5) on inside for comfort
  • Versatile protective garment for non-hazardous substances
Specifications
Specifications
Unisex
White
Elastic
Zipper
Elastic
Polyethylene
Single-use
Hooded
Non-sterile