missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Proshield™ 4 Practik
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Description
- Micro-perforated non-woven polyethylene fabric allows unrestricted passage of air and water vapour
- Certified according to Regulation (EU) 2016/425
- Chemical protective clothing, Category I
- Antistatic treatment (EN 1149-5) on inside for comfort
- Versatile protective garment for non-hazardous substances
Specifications
Specifications
Size | X-Large |
Gender | Unisex |
Material | Polyethylene |
Color | White |
Disposable | Single-use |
Cuff Style | Elastic |
Neck Style | Hooded |
Closure Type | Zipper |
Sterility | Non-sterile |
Sleeve Style | Elastic |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction