missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pregcy zone protein Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | pregcy zone protein |
---|---|
Concentration | 0.31 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16891335
|
Proteintech
21742-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16881335
|
Proteintech
21742-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
pregcy zone protein | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
P20742 | |
5858 | |
pregcy zone protein Fusion Protein Ag16471 | |
Primary | |
-20°C | |
PZP |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CPAMD6, pregcy zone protein, PZP | |
PZP | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title