missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pregcy zone protein Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21742-1-AP-150UL
This item is not returnable.
View return policy
Specifications
pregcy zone protein | |
Polyclonal | |
Unconjugated | |
PZP | |
CPAMD6, pregcy zone protein, PZP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5858 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20742 | |
PZP | |
pregcy zone protein Fusion Protein Ag16471 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction