Learn More
Potassium tetrachloroplatinate(II), 99.9% (metals basis), Pt 46.0% min, Thermo Scientific Chemicals
124.62 € - 2492.40 €
Chemical Identifiers
CAS | 10025-99-7 |
---|---|
Molecular Formula | Cl4H4K2Pt |
Molecular Weight (g/mol) | 419.11 |
MDL Number | MFCD00011378 |
InChI Key | JCAXKWJYKUEOFO-UHFFFAOYSA-N |
Synonym | potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate |
PubChem CID | 61440 |
SMILES | Cl.Cl.Cl.Cl.[K+].[K+].[Pt--] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11301698
|
Thermo Scientific Alfa Aesar
011048.03 |
1 g |
124.62 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11391688
|
Thermo Scientific Alfa Aesar
011048.06 |
5 g |
540.69 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11381688
|
Thermo Scientific Alfa Aesar
011048.14 |
25 g |
2492.40 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium tetrachloroplatinate solution is used in the preparation of platinum (Pt) nanoparticles. Ligands like ammonia or triphenyl phosphine can able to replace the chloride ligands to afford various derivatives. The replacement by ammonia results in the preparation of cisplatin, which is used in the cancer treatment. Further, it is used as catalyst in the hydroarylation reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10025-99-7 | |
419.11 | |
JCAXKWJYKUEOFO-UHFFFAOYSA-N | |
61440 |
Cl4H4K2Pt | |
MFCD00011378 | |
potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate | |
Cl.Cl.Cl.Cl.[K+].[K+].[Pt--] |
Specifications
(decomposition) | |
Red to Brown | |
99.9% (metals basis) | |
MFCD00011378 | |
Soluble in hot water. Slightly soluble in water. | |
Cl.Cl.Cl.Cl.[K+].[K+].[Pt--] | |
419.11 | |
415.11 | |
Red-brown crystals | |
Potassium tetrachloroplatinate(II), Pt 46.0% min. |
10025-99-7 | |
Crystalline | |
Cl4H4K2Pt | |
potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate | |
JCAXKWJYKUEOFO-UHFFFAOYSA-N | |
dipotassium;tetrachloroplatinum(2-) | |
61440 | |
Odorless | |
3.38 |
Safety and Handling
P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P342+P311-P362-P501c
H301-H315-H317-H318-H334-H335
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III
missing translation for 'einecsNumber' : 233-050-9
missing translation for 'rtecsNumber' : TP1850000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only