missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium tetrachloroplatinate(II), 99.9% (metals basis), Pt 46.0% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Potassium tetrachloroplatinate solution is used in the preparation of platinum (Pt) nanoparticles. Ligands like ammonia or triphenyl phosphine can able to replace the chloride ligands to afford various derivatives. The replacement by ammonia results in the preparation of cisplatin, which is used in the cancer treatment. Further, it is used as catalyst in the hydroarylation reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Red to Brown |
Physical Form | Crystalline |
Quantity | 25 g |
Assay Percent Range | 99.9% (metals basis) |
IUPAC Name | dipotassium tetrachloroplatinumdiuide |
Formula Weight | 415.11 |
Odor | Odorless |
Appearance | Red-brown crystals |
Chemical Name or Material | Potassium tetrachloroplatinate(II), Pt 46.0% min. |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'