Learn More
Potassium nitrite, 96+%, ACS reagent, Thermo Scientific Chemicals
20.90 € - 310.55 €
Chemical Identifiers
CAS | 7758-09-0 |
---|---|
Molecular Formula | KNO2 |
Molecular Weight (g/mol) | 85.10 |
MDL Number | MFCD00011408 |
InChI Key | BXNHTSHTPBPRFX-UHFFFAOYSA-M |
Synonym | potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320 |
PubChem CID | 516910 |
SMILES | [K+].[O-]N=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10597942
|
Thermo Scientific Acros
423060050 |
5 g |
20.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10697221
|
Thermo Scientific Acros
423065000 |
500 g |
96.48 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10434505
|
Thermo Scientific Acros
423060025 |
2.5 kg |
310.55 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7758-09-0 | |
85.10 | |
BXNHTSHTPBPRFX-UHFFFAOYSA-M | |
516910 |
KNO2 | |
MFCD00011408 | |
potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320 | |
[K+].[O-]N=O |
Specifications
387.0°C | |
White to Yellow | |
96+% | |
KNO2 | |
10,329 | |
potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320 | |
BXNHTSHTPBPRFX-UHFFFAOYSA-M | |
potassium;nitrite | |
516910 | |
≥96% | |
Glass bottle |
7758-09-0 | |
Crystals | |
KNO2 | |
MFCD00011408 | |
15, 7770 | |
Solubility in water: 3000g/L water (20°C). Other solubilities: 413 g/100mL water (100°C), slightly soluble in alcohol, soluble in hot alcohol, very soluble in liquid nh3 | |
[K+].[O-]N=O | |
85.10 | |
85.1 | |
ACS Reagent | |
Potassium nitrite, 96+% |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Very toxic to aquatic life.
May intensify fire; oxidizer.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-832-4
missing translation for 'rtecsNumber' : TT3750000
missing translation for 'tsca' : TSCA
- Potassium nitrite
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Oxidising solid Category 2
Hazard Statement
- H400-Very toxic to aquatic life.
- H301-Toxic if swallowed.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only