missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium nitrite, 96+%, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 387.0°C |
Color | White to Yellow |
Physical Form | Crystals |
Quantity | 2.5 kg |
Assay Percent Range | 96+% |
Linear Formula | KNO2 |
Fieser | 10,329 |
Merck Index | 15, 7770 |
Solubility Information | Solubility in water: 3000g/L water (20°C). Other solubilities: 413 g/100mL water (100°C), slightly soluble in alcohol, soluble in hot alcohol, very soluble in liquid nh3 |
IUPAC Name | potassium nitrite |
Show More |
Safety and Handling
Product Identifier
- Potassium nitrite
- Danger
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Oxidising solid Category 2
- H400-Very toxic to aquatic life.
- H301-Toxic if swallowed.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'