missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chloride, puriss. p.a., Honeywell Fluka™
Puriss. p.a.,≥99.5% (AT)
44.12 € - 336.95 €
Chemical Identifiers
CAS | 7447-40-7 |
---|---|
Molecular Formula | ClK |
Molecular Weight (g/mol) | 74.55 |
MDL Number | MFCD00011360 |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
PubChem CID | 4873 |
ChEBI | CHEBI:32588 |
SMILES | [Cl-].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15685920
|
Honeywell-Fluka
60130-250G |
250 g |
44.12 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15675920
|
Honeywell-Fluka
60130-1KG |
1 kg |
82.44 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15695920
|
Honeywell-Fluka
60130-5KG |
5 kg |
336.95 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
7447-40-7 | |
74.55 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
4873 | |
[Cl-].[K+] |
ClK | |
MFCD00011360 | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
CHEBI:32588 |
Specifications
7447-40-7 | |
KCl | |
NONH for all modes of transport | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium chloride | |
4873 | |
74.55g/mol | |
puriss. p.a. | |
Potassium chloride |
ClK | |
MFCD00011360 | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
[Cl-].[K+] | |
74.55 | |
CHEBI:32588 | |
≥99.5% (AT) | |
1.98g/mL at 25°C |
Safety and Handling
missing translation for 'rtecsNumber' : TS8050000