missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, puriss. p.a., Honeywell Fluka™

Product Code. 15695920
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a.,≥99.5% (AT)

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Quantity 5 kg
Linear Formula KCl
UN Number NONH for all modes of transport
IUPAC Name potassium;chloride
Formula Weight 74.55
Percent Purity ≥99.5% (AT)
Density 1.98g/mL at 25°C
Chemical Name or Material Potassium chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'