missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chloride, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E508, 99 to 100.5% (AT), ≤0.0001% Al, Honeywell Fluka™
Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E508, 99-100.5% (AT), ≤0.0001% Al
27.04 € - 502.32 €
Chemical Identifiers
CAS | 7447-40-7 |
---|---|
Molecular Formula | ClK |
Molecular Weight (g/mol) | 74.55 |
MDL Number | MFCD00011360 |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
PubChem CID | 4873 |
ChEBI | CHEBI:32588 |
SMILES | [Cl-].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15611170
|
Honeywell-Fluka
12636-250G |
250 g | |||||||
15641170
|
Honeywell-Fluka
12636-500G |
500 g |
56.91 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15691160
|
Honeywell-Fluka
12636-1KG |
1 kg |
41.80 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15601170
|
Honeywell-Fluka
12636-2.5KG |
2.5 kg | |||||||
15651170
|
Honeywell-Fluka
12636-5KG |
5 kg |
195.45 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15661170
|
Honeywell-Fluka
12636-6X1KG |
6 x 1 kg |
208.15 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15671170
|
Honeywell-Fluka
12636-6X2.5KG |
6 x 2.5 kg | |||||||
15631170
|
Honeywell-Fluka
12636-4X5KG |
4 x 5 kg | |||||||
15621170
|
Honeywell-Fluka
12636-25KG |
25 kg |
263.95 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7447-40-7 | |
74.55 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
4873 | |
[Cl-].[K+] |
ClK | |
MFCD00011360 | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
CHEBI:32588 |
Specifications
7447-40-7 | |
KCl | |
NONH for all modes of transport | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium chloride | |
4873 | |
74.55g/mol | |
puriss. | |
1.98g/mL at 25°C |
ClK | |
MFCD00011360 | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
[Cl-].[K+] | |
74.55 | |
CHEBI:32588 | |
99 to 100.5% (AT) | |
Plastic Bottle | |
Potassium Chloride, Meets analytical specification of Ph. Eur.BP, USP, FCC |
Safety and Handling
missing translation for 'einecsNumber' : 231-211-8
missing translation for 'rtecsNumber' : TS8050000