missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chloride, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E508, 99 to 100.5% (AT), ≤0.0001% Al, Honeywell Fluka™
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Boiling Point | 1,413°C |
Melting Point | 770°C (literature) |
CAS Max % | ≤100.0000% |
Quantity | 25 kg |
Assay Percent Range | 99 to 100.5% |
Linear Formula | KCl |
UN Number | NONH for all modes of transport |
IUPAC Name | potassium chloride |
Formula Weight | 74.55 |
Percent Purity | 99 to 100.5% (AT) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'