missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bromide, For IR spectroscopy, Honeywell Fluka™
for IR spectroscopy
186.08 € - 973.84 €
Chemical Identifiers
CAS | 7758-02-3 |
---|---|
Molecular Formula | BrK |
Molecular Weight (g/mol) | 119.00 |
MDL Number | MFCD00011358 |
InChI Key | IOLCXVTUBQKXJR-UHFFFAOYSA-M |
Synonym | potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 |
PubChem CID | 253877 |
ChEBI | CHEBI:32030 |
SMILES | [K+].[Br-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15624700
|
Honeywell-Fluka
34919-100G |
100 g |
186.08 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15644700
|
Honeywell-Fluka
34919-6X100G |
6 x 100 g |
973.84 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7758-02-3 | |
119.00 | |
IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
253877 | |
[K+].[Br-] |
BrK | |
MFCD00011358 | |
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
CHEBI:32030 |
Specifications
7758-02-3 | |
KBr | |
NONH for all modes of transport | |
IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
potassium;bromide | |
253877 | |
119g/mol | |
Potassium bromide |
BrK | |
MFCD00011358 | |
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
[K+].[Br-] | |
119.00 | |
CHEBI:32030 | |
0.01 mmHg (20°C), 1 mmHg (795°C) |
Safety and Handling
P280-P305 + P351 + P338-P337 + P313
missing translation for 'rtecsNumber' : TS7650000