missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, For IR spectroscopy, Honeywell Fluka™

Product Code. 15644700
Click to view available options
Quantity:
100 g
6 x 100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

for IR spectroscopy

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-02-3
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

Boiling Point 1,383°C
Melting Point 734°C (literature)
CAS Max % 100.0000%
Quantity 6 x 100 g
Linear Formula CH3(CH2)3CH3
IUPAC Name potassium bromide
Formula Weight 119
Grade Spectroscopy
Packaging Glass Bottle
Density 2.73 g/cm3
Chemical Name or Material Potassium bromide, For IR spectroscopy
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'