missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, ACS, 99% min, Thermo Scientific Chemicals

For Manufacturing of Photographic Papers, Process Engraving and Lithography, Spectroscopy

27.74 € - 496.47 €

Chemical Identifiers

CAS 7758-02-3
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11357467
View Documents
Thermo Scientific Alfa Aesar
012612.18
50 g
27.74 €
50g
Please to purchase this item. Need a web account? Register with us today!
11327467
View Documents
Thermo Scientific Alfa Aesar
012612.30
250 g
50.95 €
250g
Please to purchase this item. Need a web account? Register with us today!
11337467
View Documents
Thermo Scientific Alfa Aesar
012612.A1
1 kg
121.61 €
1kg
Please to purchase this item. Need a web account? Register with us today!
11347467
View Documents
Thermo Scientific Alfa Aesar
012612.A7
5 kg
496.47 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used in manufacturing of photographic papers, engraving and lithography, in spectroscopy, and as an analytical lab reagent. Potassium bromide is applied as a veterinary drug, as an antiepileptic medication for dogs. It is utilized for the bromination of alkenes using cerium(IV) ammonium nitrate. It plays an essential role with diacetoxy iodobenzene for the efficient bromination of alkenes and alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7758-02-3
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
253877
[K+].[Br-]
BrK
MFCD00011358
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
CHEBI:32030

Specifications

1435°C
7758-02-3
Crystalline
BrK
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide
253877
119.01
Odorless
Hygroscopic
2.75 g/cm3 at 20°C
730°C
White
99% min.
MFCD00011358
Freely soluble in water. Soluble in alcohol,glycerol.
[K+].[Br-]
119.00
CHEBI:32030
ACS
1.559
White powder/crystalline/beads
Potassium bromide, For analysis ACS
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c

H319-H335

missing translation for 'einecsNumber' : 231-830-3

missing translation for 'rtecsNumber' : TS7650000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Product Identifier
  • Potassium bromide

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Potassium bromide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only