Learn More
Potassium acetate, ACS, 99% min, Thermo Scientific Chemicals
31.60 € - 555.00 €
Chemical Identifiers
CAS | 127-08-2 |
---|---|
Molecular Formula | C2H3KO2 |
Molecular Weight (g/mol) | 98.142 |
MDL Number | MFCD00012458 |
InChI Key | SCVFZCLFOSHCOH-UHFFFAOYSA-M |
Synonym | potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 |
PubChem CID | 517044 |
ChEBI | CHEBI:32029 |
IUPAC Name | potassium;acetate |
SMILES | CC(=O)[O-].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356798
|
Thermo Scientific Alfa Aesar
013449.22 |
100 g |
31.60 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346798
|
Thermo Scientific Alfa Aesar
013449.36 |
500 g |
61.60 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11366798
|
Thermo Scientific Alfa Aesar
013449.A3 |
2 kg |
162.00 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376798
|
Thermo Scientific Alfa Aesar
013449.F7 |
5 x 2 kg |
555.00 €
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium acetate is used as deicer to remove snow and ice from the surface and is considered as a replacement for calcium chloride or magnesium chloride. It is used as a food additive; as a buffer in biological research; as extinguishing agent used in class K fire extinguishers and as a catalyst to produce polyurethane polymers. It is utilized for the precipitation of dodecyl sulfate bound proteins thereby removal of proteins from DNA. It plays an important role for preserving tissues, fixation and mummification. .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
127-08-2 | |
98.142 | |
SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
517044 | |
potassium;acetate |
C2H3KO2 | |
MFCD00012458 | |
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
CHEBI:32029 | |
CC(=O)[O-].[K+] |
Spécification
292°C | |
Crystalline | |
C2H3KO2 | |
3595449 | |
Soluble in water,alcohol and ammonia. Insoluble in acetone and ether. | |
CC(=O)[O-].[K+] | |
98.142 | |
CHEBI:32029 | |
ACS Reagent | |
1.57 g/mL |
127-08-2 | |
99% min | |
MFCD00012458 | |
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
potassium;acetate | |
517044 | |
98.15 | |
Hygroscopic | |
Potassium acetate, For ACS analysis |
Sécurité et traitement
H500
missing translation for 'einecsNumber' : 204-822-2
missing translation for 'rtecsNumber' : AJ3325000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only