missing translation for 'onlineSavingsMsg'
Learn More

Potassium acetate, ACS, 99% min, Thermo Scientific Chemicals

Product Code. 11366798
Click to view available options
Quantity:
100 g
500 g
2 kg
5 x 2 kg

Potassium acetate is used as deicer to remove snow and ice from the surface and is considered as a replacement for calcium chloride or magnesium chloride. It is used as a food additive; as a buffer in biological research; as extinguishing agent used in class K fire extinguishers and as a catalyst to produce polyurethane polymers. It is utilized for the precipitation of dodecyl sulfate bound proteins thereby removal of proteins from DNA. It plays an important role for preserving tissues, fixation and mummification. .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

Melting Point 292°C
Color White
Physical Form Crystalline
Quantity 2 kg
Linear Formula CH3CO2K
Beilstein 3595449
Merck Index 14,7605
Solubility Information Soluble in water,alcohol and ammonia; Insoluble in acetone and ether.
Formula Weight 98.15
Percent Purity ≥99%
Grade ACS
Sensitivity Hygroscopic
Density 1.57
Chemical Name or Material Potassium acetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'