missing translation for 'onlineSavingsMsg'
Learn More

Phospho-MARCKS (Ser93, Ser97, Ser104) Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

135.00 € - 360.00 €

Specifications

Antigen Phospho-MARCKS (Ser93, Ser97, Ser104)
Concentration 0.13 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16886163
View Documents Promotion Details
Proteintech
10018-3-AP-20UL
20 μL
135.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16876163
View Documents Promotion Details
Proteintech
10018-3-AP-100UL
100 μL
360.00 €
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody is raised against phospho-Marcks with S93, S97 and S104 phosphorylated.

MARCKS is a substrate for protein kinase C. It is localized to the plasma membrane and is an actin filament crosslinking protein. Phosphorylation by protein kinase C or binding to calcium-calmodulin inhibits its association with actin and with the plasma membrane, leading to its presence in the cytoplasm. The protein is thought to be involved in cell motility, phagocytosis, membrane trafficking and mitogenesis.
Specifications

Specifications

Phospho-MARCKS (Ser93, Ser97, Ser104)
Western Blot
Unconjugated
Rabbit
Mouse, Human
P26645, P29966
17118, 4082
Peptide
Primary
-20°C
MARCKS
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Macs, Marcks, phospho-Marcks, PKCSL
Marcks
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications