missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MARCKS (Ser93, Ser97, Ser104) Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10018-3-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is raised against phospho-Marcks with S93, S97 and S104 phosphorylated.
MARCKS is a substrate for protein kinase C. It is localized to the plasma membrane and is an actin filament crosslinking protein. Phosphorylation by protein kinase C or binding to calcium-calmodulin inhibits its association with actin and with the plasma membrane, leading to its presence in the cytoplasm. The protein is thought to be involved in cell motility, phagocytosis, membrane trafficking and mitogenesis.Specifications
Phospho-MARCKS (Ser93, Ser97, Ser104) | |
Polyclonal | |
Unconjugated | |
MARCKS | |
Macs, Marcks, phospho-Marcks, PKCSL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17118, 4082 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P26645, P29966 | |
Marcks | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |